siRNA Details
Gene Symbol: Cdc27 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001285988.1 | NP_001272917.1 | 5 | 639 | ||
NM_001285989.1 | NP_001272918.1 | 5 | 639 | |||
NM_145436.2 | NP_663411.2 | 5 | 639 | |||
GenBank mRNA | AK039377.1 | - | 5 | 436 | ||
AK134425.1 | - | 4 | 351 | |||
AK148278.1 | - | 5 | 495 | |||
AK161471.1 | - | 5 | 508 | |||
BC023187.1 | - | 5 | 497 | |||
BC157955.1 | - | 5 | 638 | 9088128 | ||
BC172100.1 | - | 5 | 638 | 9054386 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
