siRNA Details
Gene Symbol: Cstf3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_145529.3 | NP_663504.1 | 8 | 720 | ||
GenBank mRNA | AK159180.1 | - | 8 | 523 | ||
BC003241.1 | - | 8 | 681 | 3498424 |

More Information
Other Species Targeted

Gene Ontology Categories:
