siRNA Details
Gene Symbol: CKB | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001823.4 | NP_001814.2 | 3 | 351 | ||
XM_017020951.1 | XP_016876440.1 | 3 | 325 | |||
XM_017020952.1 | XP_016876441.1 | 3 | 325 | |||
GenBank mRNA | AK290101.1 | - | 3 | 322 | ||
AK312282.1 | - | 3 | 323 | |||
BC001190.1 | - | 3 | 310 | 3354679 | ||
BC004914.2 | - | 3 | 302 | 3537262 | ||
BC008323.1 | - | 3 | 308 | 3358085 | ||
BC010002.2 | - | 3 | 317 | 4135106 | ||
BC019259.1 | - | 3 | 306 | 3503057 | ||
BC019281.1 | - | 3 | 316 | 2823530 | ||
BG720434.1 | - | 3 | 333 | |||
BP213119.1 | - | 3 | 351 | |||
CR542268.1 | - | 2 | 245 | |||
EU794629.1 | - | 3 | 306 | |||
FJ224337.1 | - | 2 | 245 | |||
KU177977.1 | - | 2 | 245 | |||
KU177978.1 | - | 2 | 245 | |||
L47647.1 | - | 3 | 288 | |||
M16364.1 | - | 2 | 253 | |||
M16451.1 | - | 2 | 268 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
