siRNA Details
Gene Symbol: Larp1b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001040399.1 | NP_001035489.1 | 10 | 1326 | ||
GenBank mRNA | AK076732.1 | - | 10 | 1294 | ||
AK160155.1 | - | 10 | 1326 | |||
AK163348.1 | - | 9 | 1292 | |||
BC025528.1 | - | 8 | 1249 | 5324328 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
