siRNA Details
Gene Symbol: Tet2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001040400.2 | NP_001035490.2 | 5294 | |||
GenBank mRNA | AK129389.1 | - | 4 | 2481 | ||
AK151359.1 | - | 9 | 2284 | |||
AK152297.1 | - | 9 | 2284 | |||
AK153251.1 | - | 9 | 2284 | |||
AK153311.1 | - | 9 | 2284 | |||
AK153460.1 | - | 9 | 2284 | |||
BC031159.1 | - | 3 | 1469 | |||
BC040785.1 | - | 3 | 1469 | |||
DQ079067.1 | - | 12 | 5293 | |||
GU079949.1 | - | 10 | 4939 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
