siRNA Details
Gene Symbol: Chd3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_146019.4 | NP_666131.3 | 40 | 6906 | ||
GenBank mRNA | AK011183.1 | - | 3 | 1102 | ||
BC020471.1 | - | 3 | 1106 | |||
BC030435.1 | - | 7 | 1789 | |||
BC049913.1 | - | 3 | 1339 | |||
BC055484.1 | - | 3 | 1393 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
