siRNA Details
Gene Symbol: BC006965 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK035132.1 | - | 474 | |||
AK048256.1 | - | 2 | 407 | |||
AK145007.1 | - | 2 | 409 | |||
BC006965.1 | - | 3 | 580 | |||
BC010247.1 | - | 3 | 298 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
