siRNA Details
Gene Symbol: CNIH1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_005776.2 | NP_005767.1 | 5 | 1337 | ||
GenBank mRNA | AF070654.1 | - | 5 | 1293 | ||
AF104398.1 | - | 5 | 1297 | |||
AK074781.1 | - | 5 | 1242 | |||
AK307074.1 | - | 4 | 1098 | |||
AY358635.1 | - | 5 | 1247 | |||
BC103741.1 | - | 5 | 1338 | 6025795 | ||
BX248284.1 | - | 4 | 1086 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
