siRNA Details
Gene Symbol: Rabgap1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_146121.2 | NP_666233.2 | 23 | 3007 | ||
GenBank mRNA | AK051644.1 | - | 9 | 850 | ||
AK220489.1 | - | 22 | 2828 | |||
BC026862.1 | - | 6 | 538 | |||
BC031714.1 | - | 7 | 589 | |||
BC138337.1 | - | 23 | 2909 | 8861357 |

More Information
Other Species Targeted

Gene Ontology Categories:
