siRNA Details
Gene Symbol: Rabgap1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_146121.2 | NP_666233.2 | 23, 24 | 3029 | ||
GenBank mRNA | AK051644.1 | - | 9, 10 | 872 | ||
AK220489.1 | - | 22, 23 | 2850 | |||
BC026862.1 | - | 6, 7 | 560 | |||
BC031714.1 | - | 7, 8 | 611 | |||
BC138337.1 | - | 23, 24 | 2931 | 8861357 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
