siRNA Details
Gene Symbol: Rabgap1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_146121.2 | NP_666233.2 | 24 | 3065 | ||
GenBank mRNA | AK051644.1 | - | 10 | 908 | ||
AK220489.1 | - | 23 | 2886 | |||
BC026862.1 | - | 7 | 596 | |||
BC031714.1 | - | 8 | 647 | |||
BC138337.1 | - | 24 | 2967 | 8861357 |

More Information
Other Species Targeted

Gene Ontology Categories:
