siRNA Details
Gene Symbol: Wdr62 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_146186.3 | NP_666298.3 | 8 | 1068 | ||
GenBank mRNA | AK154592.1 | - | 8 | 1039 | ||
AK203944.1 | - | 2 | 190 | |||
BC054747.1 | - | 8 | 1043 | 6405269 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
