siRNA Details
Gene Symbol: Wdr62 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_146186.3 | NP_666298.3 | 9, 10 | 1330 | ||
GenBank mRNA | AK154592.1 | - | 9, 10 | 1301 | ||
BC054747.1 | - | 9, 10 | 1305 | 6405269 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
