siRNA Details
Gene Symbol: Spns2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001276383.1 | NP_001263312.1 | 13 | 2485 | ||
NM_153060.3 | NP_694700.2 | 13 | 2833 | |||
GenBank mRNA | AK156007.1 | - | 14 | 2479 | ||
BC011467.1 | - | 13 | 2833 | |||
BC025823.1 | - | 3 | 1945 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
