siRNA Details
Gene Symbol: PEAK1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_024776.3 | NP_079052.2 | 8 | 9774 | ||
XM_005254670.2 | XP_005254727.1 | 11 | 10078 | |||
XM_005254671.2 | XP_005254728.1 | 10 | 9975 | |||
XM_005254673.2 | XP_005254730.1 | 8 | 9709 | |||
XM_005254674.2 | XP_005254731.1 | 9 | 9922 | |||
XM_005254675.4 | XP_005254732.1 | 5 | 9428 | |||
XM_011522033.2 | XP_011520335.1 | 12 | 10134 | |||
XM_011522034.2 | XP_011520336.1 | 13 | 10242 | |||
XM_011522035.2 | XP_011520337.1 | 15 | 10393 | |||
XM_011522036.2 | XP_011520338.1 | 14 | 10297 | |||
XM_011522037.2 | XP_011520339.1 | 14 | 10338 | |||
XM_011522038.2 | XP_011520340.1 | 13 | 10189 | |||
XM_011522039.2 | XP_011520341.1 | 14 | 10285 | |||
XM_011522042.2 | XP_011520344.1 | 4 | 6511 | |||
GenBank mRNA | AB082533.1 | - | 4 | 6247 | ||
AK124548.1 | - | 1 | 1610 |
Gene Symbol: CBFA2T2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001032999.2 | NP_001028171.1 | 11 | 4060 | ||
NM_001039709.1 | NP_001034798.1 | 11 | 4190 | |||
NM_005093.3 | NP_005084.1 | 12 | 4478 | |||
XM_011529101.2 | XP_011527403.1 | 11 | 4109 | |||
XM_011529102.2 | XP_011527404.1 | 11 | 3924 | |||
XM_011529103.2 | XP_011527405.1 | 11 | 4009 | |||
XM_011529107.2 | XP_011527409.1 | 9 | 3576 | |||
XM_017028121.1 | XP_016883610.1 | 11 | 3927 | |||
XM_017028122.1 | XP_016883611.1 | 11 | 4111 | |||
XM_017028124.1 | XP_016883613.1 | 10 | 4336 | |||
XM_017028125.1 | XP_016883614.1 | 9 | 3573 |
Gene Symbol: MAP3K11 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK092015.1 | - | 1 | 599 |
Gene Symbol: PLIN3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001164189.1 | NP_001157661.1 | 8 | 1984 | ||
NM_001164194.1 | NP_001157666.1 | 8 | 1951 | |||
NM_005817.4 | NP_005808.3 | 8 | 1987 | |||
GenBank mRNA | AF051314.1 | - | 4 | 1300 | ||
AF051315.1 | - | 4 | 1275 | |||
AF055574.1 | - | 8 | 1863 | |||
AF057140.1 | - | 8 | 1885 | |||
AK226088.1 | - | 8 | 1865 | |||
BC001590.2 | - | 8 | 1860 | 2987965 | ||
BC005818.2 | - | 8 | 1876 | 3833411 | ||
BC007566.1 | - | 8 | 1877 | 3028104 | ||
BC019278.1 | - | 8 | 1876 | 2905275 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
