siRNA Details
Gene Symbol: Ppp1r16b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001159662.1 | NP_001153134.1 | 11 | 2910 | ||
NM_153089.4 | NP_694729.1 | 11 | 2888 | |||
GenBank mRNA | AK037627.1 | - | 3206 | |||
AK042123.1 | - | 11 | 2884 | |||
BC054764.1 | - | 11 | 2909 | 6414610 | ||
BC057542.1 | - | 11 | 2909 | 6414610 |
