siRNA Details
Gene Symbol: Rab11fip3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001162868.1 | NP_001156340.1 | 9 | 2483 | ||
NM_001162869.1 | NP_001156341.1 | 10 | 2618 | |||
NM_153140.2 | NP_694780.1 | 7 | 874 | |||
GenBank mRNA | AB093257.1 | - | 10 | 2059 | ||
AK083658.1 | - | 9 | 2167 | |||
AK085516.1 | - | 7 | 875 | |||
AK128985.1 | - | 7 | 874 | |||
BC023364.1 | - | 7 | 865 | |||
BC037132.1 | - | 7 | 914 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
