siRNA Details
Gene Symbol: Rab11fip3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001162868.1 | NP_001156340.1 | 12 | 2765 | ||
NM_001162869.1 | NP_001156341.1 | 13 | 2900 | |||
NM_153140.2 | NP_694780.1 | 10 | 1156 | |||
GenBank mRNA | AB093257.1 | - | 13 | 2341 | ||
AK083658.1 | - | 12 | 2450 | |||
AK085516.1 | - | 10 | 1157 | |||
AK128985.1 | - | 10 | 1156 | |||
BC023364.1 | - | 10 | 1147 | |||
BC037132.1 | - | 10 | 1196 | |||
CB840427.1 | - | 1 | 51 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
