siRNA Details
Gene Symbol: Rab11fip3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001162868.1 | NP_001156340.1 | 14 | 4459 | ||
NM_001162869.1 | NP_001156341.1 | 15 | 4594 | |||
NM_153140.2 | NP_694780.1 | 12 | 2850 | |||
GenBank mRNA | AB093257.1 | - | 15 | 4035 | ||
AK083658.1 | - | 14 | 4148 | |||
AK085516.1 | - | 11 | 2890 | |||
AK128985.1 | - | 12 | 2850 | |||
BC023364.1 | - | 12 | 2841 | |||
BC037132.1 | - | 12 | 2890 |

More Information
Other Species Targeted

Gene Ontology Categories:
