siRNA Details
Gene Symbol: E130308A19Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_153158.5 | NP_694798.1 | 3 | 1704 | ||
GenBank mRNA | AK047601.1 | - | 3 | 1674 | ||
BC037070.1 | - | 3 | 1522 | 5004205 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
