siRNA Details
Gene Symbol: AK8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001317958.1 | NP_001304887.1 | 12, 13 | 1773 | ||
NM_001317959.1 | NP_001304888.1 | 14, 15 | 2159 | |||
NM_152572.2 | NP_689785.1 | 12, 13 | 1766 | |||
XM_005272169.2 | XP_005272226.1 | 12, 13 | 1305 | |||
XM_006716965.2 | XP_006717028.1 | 10, 11 | 1078 | |||
XM_011518278.2 | XP_011516580.1 | 11, 12 | 1146 | |||
XM_017014308.1 | XP_016869797.1 | 6, 7 | 828 | |||
GenBank mRNA | AK057266.1 | - | 12, 13 | 1239 | ||
AK093446.1 | - | 12, 13 | 1281 | |||
AK292186.1 | - | 12, 13 | 1251 | |||
BC034776.1 | - | 12, 13 | 1263 | 5175757 | ||
BC050576.1 | - | 12, 13 | 1766 | 5744517 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
