siRNA Details
Gene Symbol: GLYAT | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_005838.3 | NP_005829.3 | 5 | 959 | ||
GenBank mRNA | AF023466.1 | - | 5 | 886 | ||
BC009785.1 | - | 5 | 963 | 4045672 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
