siRNA Details
Gene Symbol: Marf1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001081154.2 | NP_001074623.1 | 5 | 1357 | ||
GenBank mRNA | AK035704.1 | - | 1011 | |||
AK220341.1 | - | 3 | 3139 | |||
BC069852.1 | - | 1 | 43 |

More Information
Other Species Targeted

Gene Ontology Categories:
