siRNA Details
Gene Symbol: Marf1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001081154.2 | NP_001074623.1 | 8 | 1890 | ||
GenBank mRNA | AK035704.1 | - | 1544 | |||
AK220341.1 | - | 5 | 3554 | |||
BC031575.1 | - | 1 | 62 | |||
BC069852.1 | - | 4 | 576 |

More Information
Other Species Targeted

Gene Ontology Categories:
