siRNA Details
Gene Symbol: STUB1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001293197.1 | NP_001280126.1 | 7 | 1376 | ||
NM_005861.3 | NP_005852.2 | 7 | 1287 | |||
GenBank mRNA | AF039689.1 | - | 7 | 928 | ||
AF129085.1 | - | 7 | 968 | |||
AF217968.1 | - | 7 | 1263 | |||
AF432221.1 | - | 7 | 983 | |||
BC007545.1 | - | 7 | 971 | 2959735 | ||
BC017178.1 | - | 7 | 958 | 3346811 | ||
BC022788.1 | - | 7 | 971 | 2959735 | ||
BC063617.1 | - | 7 | 902 | 3847168 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
