siRNA Details
Gene Symbol: Ppat | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_172146.2 | NP_742158.1 | 3 | 760 | ||
GenBank mRNA | AK147750.1 | - | 3 | 410 | ||
AK148034.1 | - | 3 | 364 | |||
AK159141.1 | - | 4 | 910 | |||
AK159859.1 | - | 4 | 910 | |||
AK160542.1 | - | 3 | 385 | |||
AK164506.1 | - | 3 | 374 | |||
AK167057.1 | - | 3 | 399 | |||
BC006053.1 | - | 3 | 347 | |||
BC023841.1 | - | 3 | 387 | 5346105 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
