siRNA Details
Gene Symbol: Megf6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001162977.1 | NP_001156449.1 | 37 | 5342 | ||
GenBank mRNA | AK134303.1 | - | 24 | 3336 | ||
AK173032.1 | - | 20 | 2821 | |||
BC031402.1 | - | 17 | 2380 | |||
BC039980.1 | - | 17 | 2391 | |||
BC058571.1 | - | 16 | 2284 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
