siRNA Details
Gene Symbol: Adamtsl1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_029967.3 | NP_084243.3 | 20 | 3910 | ||
GenBank mRNA | AK031246.1 | - | 3 | 513 | ||
AK132420.1 | - | 8 | 2350 | |||
BC116295.1 | - | 3 | 504 | |||
BC116296.1 | - | 3 | 504 |
