siRNA Details
Gene Symbol: Adamtsl1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_029967.3 | NP_084243.3 | 28 | 6894 | ||
GenBank mRNA | AK031246.1 | - | 11 | 3495 | ||
AK132420.1 | - | 16 | 5333 | |||
AK136721.1 | - | 1 | 1075 | |||
AK140099.1 | - | 1 | 1076 |
