siRNA Details
Gene Symbol: Tmem63c | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_172583.2 | NP_766171.1 | 11 | 981 | ||
GenBank mRNA | AK030062.1 | - | 11 | 973 | ||
AK034410.1 | - | 10 | 904 | |||
AK043536.1 | - | 10 | 902 | |||
BC056936.1 | - | 11 | 981 | 5695989 | ||
BC057088.1 | - | 11 | 981 | 5695989 |
