siRNA Details
Gene Symbol: A630038E17Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK040154.1 | - | 6 | 692 | ||
AK041790.1 | - | 7 | 1133 | |||
AK088167.1 | - | 6 | 791 | |||
AK147195.1 | - | 5 | 659 | |||
AK153587.1 | - | 5 | 859 | |||
BC038285.1 | - | 6 | 838 | |||
BC111819.1 | - | 7 | 1079 | |||
M37694.1 | - | 6 | 823 | |||
M37695.1 | - | 4 | 622 | |||
X12729.1 | - | 999 |
