siRNA Details
Gene Symbol: 1700021K19Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001200038.1 | NP_001186967.1 | 2 | 318 | ||
NM_172615.4 | NP_766203.1 | 2 | 318 | |||
GenBank mRNA | AK030368.1 | - | 2 | 292 | ||
AK042428.1 | - | 2 | 256 | |||
AK122221.1 | - | 2 | 282 | |||
AK170002.1 | - | 2 | 298 | |||
AK170223.1 | - | 2 | 256 | |||
AK170312.1 | - | 2 | 549 | |||
AK171583.1 | - | 2 | 302 | |||
BC057307.1 | - | 2 | 222 | |||
BC060601.1 | - | 2 | 291 | 5706123 | ||
BC067390.1 | - | 2 | 303 | 30432960 |
