siRNA Details
Gene Symbol: Treml4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001033922.2 | NP_001029094.1 | 6 | 1645 | ||
NM_001163795.1 | NP_001157267.1 | 6 | 1660 | |||
NM_001163796.1 | NP_001157268.1 | 6 | 1633 | |||
NM_172623.2 | NP_766211.2 | 6 | 1657 | |||
GenBank mRNA | AK077228.1 | - | 6 | 1657 | ||
AK156085.1 | - | 6 | 1660 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
