siRNA Details
Gene Symbol: MAT2A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_005911.5 | NP_005902.1 | 1, 2 | 400 | ||
GenBank mRNA | AK291126.1 | - | 1, 2 | 208 | ||
AK293841.1 | - | 1, 2 | 210 | |||
AK297758.1 | - | 1, 2 | 103 | |||
AK297797.1 | - | 1, 2 | 210 | |||
AK308591.1 | - | 1, 2 | 103 | |||
BC001686.2 | - | 1, 2 | 197 | 3010820 | ||
BC001854.2 | - | 1, 2 | 197 | 3010820 | ||
BI549760.1 | - | 1, 2 | 219 | |||
DC322518.1 | - | 1, 2 | 241 | |||
DQ083239.1 | - | 1, 2 | 136 | |||
X68836.1 | - | 1, 2 | 155 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
