siRNA Details
Gene Symbol: Fam178a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001081225.1 | NP_001074694.1 | 11 | 2798 | ||
GenBank mRNA | AK028324.1 | - | 7 | 749 | ||
AK031688.1 | - | 11 | 2770 | |||
AK032405.1 | - | 7 | 753 | |||
AK036700.1 | - | 9 | 2375 | |||
AK147588.1 | - | 11 | 3271 | |||
AK161809.1 | - | 7 | 1161 | |||
BC060679.1 | - | 7 | 715 | |||
BC150999.1 | - | 11 | 2651 | 9087912 | ||
BC157937.1 | - | 11 | 2651 | 9088117 | ||
BC172097.1 | - | 11 | 2651 | 9054383 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
