siRNA Details
Gene Symbol: Suco | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_172645.2 | NP_766233.2 | 8, 9 | 1282 | ||
GenBank mRNA | AK029097.1 | - | 2, 3 | 271 | ||
AK171856.1 | - | 8, 9 | 1221 | |||
BC138379.1 | - | 8, 9 | 1201 | 8861399 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
