siRNA Details
Gene Symbol: Suco | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_172645.2 | NP_766233.2 | 18 | 3067 | ||
GenBank mRNA | AK029097.1 | - | 12 | 2056 | ||
BC023426.1 | - | 6 | 1460 | |||
BC138379.1 | - | 18 | 2986 | 8861399 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
