siRNA Details
Gene Symbol: 4932438A13Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_172679.2 | NP_766267.2 | 11 | 1392 | ||
GenBank mRNA | AK028716.1 | - | 10 | 1068 | ||
AK030086.1 | - | 11 | 1379 | |||
AK148181.1 | - | 8 | 902 | |||
AK220265.1 | - | 11 | 1436 | |||
BC034738.1 | - | 3 | 241 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
