siRNA Details
Gene Symbol: 4932438A13Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_172679.2 | NP_766267.2 | 76 | 13403 | ||
GenBank mRNA | AK041642.1 | - | 3 | 487 | ||
AK052702.1 | - | 8 | 1221 | |||
AK088301.1 | - | 1 | 992 | |||
AK164468.1 | - | 2 | 1001 | |||
AK173100.1 | - | 20 | 3038 | |||
BC027125.1 | - | 3 | 435 | |||
BC079623.1 | - | 17 | 2773 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
