siRNA Details
Gene Symbol: 4932438A13Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_172679.2 | NP_766267.2 | 82, 83 | 14687 | ||
GenBank mRNA | AK041642.1 | - | 9, 10 | 1771 | ||
AK052702.1 | - | 14, 15 | 2505 | |||
AK134811.1 | - | 3, 4 | 657 | |||
AK173100.1 | - | 26, 27 | 4322 | |||
BC024954.1 | - | 4, 5 | 801 | |||
BC027125.1 | - | 9, 10 | 1719 | |||
BC079623.1 | - | 23, 24 | 4057 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
