siRNA Details
Gene Symbol: Eif4g3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001256195.1 | NP_001243124.1 | 26, 27 | 4156 | ||
NM_001256198.1 | NP_001243127.1 | 26, 27 | 4213 | |||
NM_172703.3 | NP_766291.2 | 26, 27 | 4060 | |||
GenBank mRNA | AK029440.1 | - | 9, 10 | 1140 | ||
AK133389.1 | - | 6, 7 | 624 | |||
AK137712.1 | - | 2, 3 | 282 | |||
BC023898.1 | - | 13, 14 | 1902 | |||
BC047531.1 | - | 15, 16 | 2284 | |||
BC048848.1 | - | 5, 6 | 566 | |||
BC057913.1 | - | 6, 7 | 682 | |||
BC072600.1 | - | 26, 27 | 4046 | 6410320 | ||
BC075705.1 | - | 11, 12 | 1626 | |||
BC079885.1 | - | 26, 27 | 4110 | |||
BC082549.1 | - | 26, 27 | 4177 |
