siRNA Details
Gene Symbol: Tmem87a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001110496.1 | NP_001103966.1 | 20 | 1728 | ||
NM_001110497.1 | NP_001103967.1 | 20 | 1725 | |||
NM_173734.3 | NP_776095.2 | 20 | 1731 | |||
GenBank mRNA | AK044591.1 | - | 20 | 1682 | ||
AK086738.1 | - | 20 | 1675 | |||
AK088559.1 | - | 19 | 1636 | |||
AK168093.1 | - | 1 | 61 | |||
BC004047.1 | - | 20 | 1677 | |||
BC024676.1 | - | 10 | 679 | |||
BC027354.1 | - | 20 | 1704 | 4010477 |
