siRNA Details
Gene Symbol: CCT3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_005998.4 | NP_005989.3 | 4 | 405 | ||
NR_036564.1 | 5 | 567 | ||||
NR_036565.1 | 5 | 518 | ||||
GenBank mRNA | AB208882.1 | - | 4 | 271 | ||
AK126400.1 | - | 4 | 261 | |||
AK300765.1 | - | 5 | 399 | |||
AK308023.1 | - | 4 | 282 | |||
AK315923.1 | - | 3 | 191 | |||
AK316387.1 | - | 5 | 310 | |||
AL833197.1 | - | 4 | 265 | |||
BC006501.2 | - | 4 | 237 | 2820063 | ||
BC008019.1 | - | 4 | 247 | 3534941 | ||
BG476176.1 | - | 4 | 406 | |||
BG746513.1 | - | 4 | 220 | |||
X74801.1 | - | 4 | 171 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
