siRNA Details
Gene Symbol: Mmgt2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_175002.2 | NP_778167.1 | 2 | 697 | ||
GenBank mRNA | AK004099.1 | - | 2 | 534 | ||
AK131977.1 | - | 2 | 587 | |||
BC025076.1 | - | 1 | 468 | 4459181 | ||
BC096408.1 | - | 3 | 312 | |||
BC096583.1 | - | 2 | 641 | 5025927 | ||
EU069462.1 | - | 1 | 355 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
