siRNA Details
Gene Symbol: Mmgt2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_175002.2 | NP_778167.1 | 2 | 765 | ||
GenBank mRNA | AK131977.1 | - | 2 | 655 | ||
BC025076.1 | - | 1 | 536 | 4459181 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
