siRNA Details
Gene Symbol: Wdr18 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK032251.1 | - | 2 | 722 | ||
AK087505.1 | - | 1 | 1355 | |||
AK163465.1 | - | 1 | 1352 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
