siRNA Details
Gene Symbol: Gpr179 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001081220.1 | NP_001074689.1 | 1 | 273 | ||
GenBank mRNA | AK030625.1 | - | 1 | 277 | ||
AK038894.1 | - | 1 | 267 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
