siRNA Details
Gene Symbol: Glis3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK054020.1 | - | 4 | 1701 | ||
AY220846.1 | - | 2 | 746 | |||
BC066143.1 | - | 3 | 1081 | |||
BC129906.1 | - | 2 | 724 | |||
BC129907.1 | - | 2 | 724 | |||
DQ871027.1 | - | 4 | 1701 |
