siRNA Details
Gene Symbol: Kcnt1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK033953.1 | - | 6, 7 | 502 | ||
AK086119.1 | - | 5, 6 | 474 | |||
AK147625.1 | - | 4, 5 | 2451 | |||
BB625805.1 | - | 6, 7 | 494 | |||
BB662543.1 | - | 5, 6 | 469 | |||
BC141190.1 | - | 5, 6 | 547 | 9055802 | ||
BC171963.1 | - | 5, 6 | 547 | 9054249 | ||
FJ628361.1 | - | 6, 7 | 545 |

More Information
Other Species Targeted

