siRNA Details
Gene Symbol: A930012O16Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK044432.1 | - | 1210 | |||
AK050290.1 | - | 2 | 927 | |||
AK169486.1 | - | 1780 |

UserName
Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: A930012O16Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK044432.1 | - | 1210 | |||
AK050290.1 | - | 2 | 927 | |||
AK169486.1 | - | 1780 |